siRNA Details
Gene Symbol: Fscn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100806.1 | NP_001094276.1 | 3 | 1307 | ||
GenBank mRNA | AY268934.1 | - | 141 | |||
BC168678.1 | - | 3 | 1307 | 5598674 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fscn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100806.1 | NP_001094276.1 | 3 | 1307 | ||
GenBank mRNA | AY268934.1 | - | 141 | |||
BC168678.1 | - | 3 | 1307 | 5598674 |