siRNA Details
| Gene Symbol: LOC683897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109448.1 | NP_001102918.1 | 2 | 207 | ||
| XM_006256581.2 | XP_006256643.1 | 3 | 254 | |||
| GenBank mRNA | BC105890.1 | - | 1 | 136 | 7306492 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC683897| Gene Symbol: LOC683897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109448.1 | NP_001102918.1 | 2 | 207 | ||
| XM_006256581.2 | XP_006256643.1 | 3 | 254 | |||
| GenBank mRNA | BC105890.1 | - | 1 | 136 | 7306492 | |