siRNA Details
Gene Symbol: Pwwp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246135.2 | XP_006246197.1 | 2 | 3097 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pwwp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246135.2 | XP_006246197.1 | 2 | 3097 |