siRNA Details
| Gene Symbol: LOC102548917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252956.2 | XP_006253018.1 | 4 | 1492 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102548917| Gene Symbol: LOC102548917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252956.2 | XP_006253018.1 | 4 | 1492 | ||