siRNA Details
| Gene Symbol: PCOLCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127640.1 | NP_001121112.1 | 7 | 1334 | ||
| GenBank mRNA | BC161891.1 | - | 7 | 1334 | 7189256 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCOLCE2| Gene Symbol: PCOLCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127640.1 | NP_001121112.1 | 7 | 1334 | ||
| GenBank mRNA | BC161891.1 | - | 7 | 1334 | 7189256 | |