siRNA Details
| Gene Symbol: Nr2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113197.1 | NP_001106668.1 | 7 | 945 | ||
| GenBank mRNA | EU316216.1 | - | 7 | 945 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr2e1| Gene Symbol: Nr2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113197.1 | NP_001106668.1 | 7 | 945 | ||
| GenBank mRNA | EU316216.1 | - | 7 | 945 | ||