siRNA Details
| Gene Symbol: Snx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044283.1 | NP_001037748.1 | 3 | 650 | ||
| GenBank mRNA | BC086341.1 | - | 3 | 414 | 7301779 | |
| BC107918.1 | - | 3 | 650 | 7458016 | ||
| BC158608.1 | - | 3 | 374 | 7301775 | ||
| FQ211490.1 | - | 3 | 355 | |||
| FQ231492.1 | - | 3 | 397 | |||
| FQ234595.1 | - | 3 | 397 | |||

Snx3
Genomic Map