siRNA Details
Gene Symbol: Rd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250496.2 | XP_006250558.1 | 2 | 218 | ||
XM_006250497.2 | XP_006250559.1 | 2 | 616 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250496.2 | XP_006250558.1 | 2 | 218 | ||
XM_006250497.2 | XP_006250559.1 | 2 | 616 |