siRNA Details
| Gene Symbol: LOC684208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748936.3 | XP_003748984.1 | 1 | 681 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC684208| Gene Symbol: LOC684208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748936.3 | XP_003748984.1 | 1 | 681 | ||