siRNA Details
| Gene Symbol: Rbm15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749380.3 | XP_003749428.1 | 1 | 3240 | ||
| XM_006233197.2 | XP_006233259.1 | 1 | 3241 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm15| Gene Symbol: Rbm15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749380.3 | XP_003749428.1 | 1 | 3240 | ||
| XM_006233197.2 | XP_006233259.1 | 1 | 3241 | |||