siRNA Details
| Gene Symbol: Mrpl39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768604.1 | XP_008766826.1 | 2 | 174 | ||
| GenBank mRNA | FQ225027.1 | - | 2 | 133 | ||
| FQ230552.1 | - | 2 | 133 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl39| Gene Symbol: Mrpl39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768604.1 | XP_008766826.1 | 2 | 174 | ||
| GenBank mRNA | FQ225027.1 | - | 2 | 133 | ||
| FQ230552.1 | - | 2 | 133 | |||