siRNA Details
| Gene Symbol: Kcmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128192.1 | NP_001121664.1 | 5 | 774 | ||
| GenBank mRNA | BC167075.1 | - | 5 | 774 | 9093013 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcmf1| Gene Symbol: Kcmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128192.1 | NP_001121664.1 | 5 | 774 | ||
| GenBank mRNA | BC167075.1 | - | 5 | 774 | 9093013 | |