siRNA Details
Gene Symbol: Epm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276762.1 | NP_001263691.1 | 2 | 323 | ||
GenBank mRNA | AF347030.1 | - | 311 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Epm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276762.1 | NP_001263691.1 | 2 | 323 | ||
GenBank mRNA | AF347030.1 | - | 311 |