siRNA Details
| Gene Symbol: LOC684471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750810.3 | XP_003750858.3 | 2 | 1919 | ||
| XM_008767723.1 | XP_008765945.1 | 2 | 2440 | |||
| XM_008767724.1 | XP_008765946.1 | 3 | 1996 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC684471| Gene Symbol: LOC684471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750810.3 | XP_003750858.3 | 2 | 1919 | ||
| XM_008767723.1 | XP_008765945.1 | 2 | 2440 | |||
| XM_008767724.1 | XP_008765946.1 | 3 | 1996 | |||