siRNA Details
| Gene Symbol: Gba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127639.1 | NP_001121111.1 | 6 | 780 | ||
| GenBank mRNA | BC166732.1 | - | 6 | 780 | 7934870 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gba| Gene Symbol: Gba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127639.1 | NP_001121111.1 | 6 | 780 | ||
| GenBank mRNA | BC166732.1 | - | 6 | 780 | 7934870 | |