siRNA Details
| Gene Symbol: Cdca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115042.2 | NP_001108514.2 | 2 | 171 | ||
| XM_008760176.1 | XP_008758398.1 | 2 | 166 | |||
| GenBank mRNA | BC097411.1 | - | 2 | 101 | ||
| BC158663.1 | - | 2 | 171 | 8369770 | ||
| BC158764.1 | - | 2 | 110 | 7111047 | ||

Cdca5
Genomic Map