siRNA Details
| Gene Symbol: Prr33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001064406.5 | XP_001064406.1 | 2 | 1977 | ||
| XM_008760181.1 | XP_008758403.1 | 2 | 1963 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr33| Gene Symbol: Prr33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001064406.5 | XP_001064406.1 | 2 | 1977 | ||
| XM_008760181.1 | XP_008758403.1 | 2 | 1963 | |||