siRNA Details
| Gene Symbol: Ces2j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190380.1 | NP_001177309.1 | 1053 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ces2j| Gene Symbol: Ces2j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190380.1 | NP_001177309.1 | 1053 | |||