siRNA Details
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031764.3 | NP_113952.2 | 7 | 1033 | ||
| XM_006250226.2 | XP_006250288.1 | 8 | 4330 | |||
| XM_006250227.2 | XP_006250289.1 | 10 | 1665 | |||
| XM_006250228.2 | XP_006250290.1 | 9 | 1446 | |||
| XM_006250229.2 | XP_006250291.1 | 9 | 1394 | |||
| XM_008769761.1 | XP_008767983.1 | 9 | 1408 | |||
| XM_008769762.1 | XP_008767984.1 | 9 | 1340 | |||
| XM_008769763.1 | XP_008767985.1 | 9 | 1646 | |||
| GenBank mRNA | BC161961.1 | - | 7 | 1033 | 9027893 | |
| EV770183.1 | - | 4 | 539 | |||

Ddr2
Genomic Map