siRNA Details
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031764.3 | NP_113952.2 | 14 | 2152 | ||
| XM_006250226.2 | XP_006250288.1 | 15 | 5449 | |||
| XM_006250227.2 | XP_006250289.1 | 17 | 2784 | |||
| XM_006250228.2 | XP_006250290.1 | 16 | 2565 | |||
| XM_006250229.2 | XP_006250291.1 | 16 | 2513 | |||
| XM_008769761.1 | XP_008767983.1 | 16 | 2527 | |||
| XM_008769762.1 | XP_008767984.1 | 16 | 2459 | |||
| XM_008769763.1 | XP_008767985.1 | 16 | 2765 | |||
| GenBank mRNA | AF016247.1 | - | 1 | 121 | ||
| BC161961.1 | - | 14 | 2152 | 9027893 | ||
| EV765307.1 | - | 3 | 398 | |||

Ddr2
Genomic Map