siRNA Details
Gene Symbol: Ttc39c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077231.1 | NP_001070699.1 | 11 | 1372 | ||
XM_006254423.2 | XP_006254485.1 | 11 | 2130 | |||
XM_008771954.1 | XP_008770176.1 | 10 | 1228 | |||
GenBank mRNA | BC105623.1 | - | 11 | 1372 | 7455888 |
