siRNA Details
| Gene Symbol: Ttc39c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077231.1 | NP_001070699.1 | 3 | 231 | ||
| XM_006254423.2 | XP_006254485.1 | 3 | 989 | |||
| XM_008771954.1 | XP_008770176.1 | 2 | 115 | |||
| GenBank mRNA | BC105623.1 | - | 3 | 231 | 7455888 | |

Ttc39c
Genomic Map