siRNA Details
Gene Symbol: Col6a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766594.1 | XP_008764816.1 | 35 | 6354 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Col6a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766594.1 | XP_008764816.1 | 35 | 6354 |