siRNA Details
| Gene Symbol: Col26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271268.1 | NP_001258197.1 | 3 | 495 | ||
| XM_008769154.1 | XP_008767376.1 | 3 | 532 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col26a1| Gene Symbol: Col26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271268.1 | NP_001258197.1 | 3 | 495 | ||
| XM_008769154.1 | XP_008767376.1 | 3 | 532 | |||