siRNA Details
Gene Symbol: Lyrm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134729.1 | NP_001128201.1 | 2, 3 | 107 | ||
GenBank mRNA | BC168191.1 | - | 2, 3 | 107 | 7107126 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lyrm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134729.1 | NP_001128201.1 | 2, 3 | 107 | ||
GenBank mRNA | BC168191.1 | - | 2, 3 | 107 | 7107126 |