siRNA Details
| Gene Symbol: Islr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126300.1 | NP_001119772.1 | 2 | 1249 | ||
| XM_006243176.2 | XP_006243238.1 | 2 | 1407 | |||
| GenBank mRNA | BC160841.1 | - | 2 | 1249 | 7461347 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Islr| Gene Symbol: Islr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126300.1 | NP_001119772.1 | 2 | 1249 | ||
| XM_006243176.2 | XP_006243238.1 | 2 | 1407 | |||
| GenBank mRNA | BC160841.1 | - | 2 | 1249 | 7461347 | |