siRNA Details
Gene Symbol: LOC100910729 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750524.3 | XP_003750572.2 | 3 | 1238 | ||
XM_006243178.2 | XP_006243240.1 | 3 | 1200 | |||
XM_006243179.2 | XP_006243241.1 | 3 | 2427 | |||
XM_008766371.1 | XP_008764593.1 | 3 | 1750 | |||
XM_008766372.1 | XP_008764594.1 | 4 | 2636 | |||
XM_008766373.1 | XP_008764595.1 | 3 | 2389 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
