siRNA Details
| Gene Symbol: Tbc1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770930.1 | XP_008769152.1 | 1 | 648 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d4| Gene Symbol: Tbc1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770930.1 | XP_008769152.1 | 1 | 648 | ||