siRNA Details
| Gene Symbol: Iqsec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236890.2 | XP_006236952.1 | 13, 14 | 3438 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001134383, AK220355, BC026481.

Iqsec1
Genomic Map