siRNA Details
| Gene Symbol: Muc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766031.1 | XP_008764253.1 | 4 | 18306 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Muc16| Gene Symbol: Muc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766031.1 | XP_008764253.1 | 4 | 18306 | ||