siRNA Details
| Gene Symbol: Ppp1r13l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228495.2 | XP_006228557.1 | 11 | 4979 | ||
| XM_008759052.1 | XP_008757274.1 | 12 | 4514 | |||
| XM_008759053.1 | XP_008757275.1 | 11 | 3683 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r13l| Gene Symbol: Ppp1r13l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228495.2 | XP_006228557.1 | 11 | 4979 | ||
| XM_008759052.1 | XP_008757274.1 | 12 | 4514 | |||
| XM_008759053.1 | XP_008757275.1 | 11 | 3683 | |||