siRNA Details
| Gene Symbol: Gstt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109675.1 | NP_001103145.1 | 3, 4 | 444 | ||
| GenBank mRNA | BC097423.1 | - | 3, 4 | 428 | 7455039 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gstt4| Gene Symbol: Gstt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109675.1 | NP_001103145.1 | 3, 4 | 444 | ||
| GenBank mRNA | BC097423.1 | - | 3, 4 | 428 | 7455039 | |