siRNA Details
| Gene Symbol: Mphosph6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129881.2 | NP_001123353.1 | 2 | 134 | ||
| GenBank mRNA | BC166883.1 | - | 2 | 134 | 6919704 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mphosph6| Gene Symbol: Mphosph6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129881.2 | NP_001123353.1 | 2 | 134 | ||
| GenBank mRNA | BC166883.1 | - | 2 | 134 | 6919704 | |