siRNA Details
| Gene Symbol: Ajap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101014.1 | NP_001094484.1 | 4 | 1335 | ||
| GenBank mRNA | AB179740.1 | - | 4 | 1335 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ajap1| Gene Symbol: Ajap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101014.1 | NP_001094484.1 | 4 | 1335 | ||
| GenBank mRNA | AB179740.1 | - | 4 | 1335 | ||