siRNA Details
| Gene Symbol: Tprg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110318.2 | NP_001103788.1 | 3 | 668 | ||
| GenBank mRNA | BC160865.1 | - | 3 | 517 | 7324838 | |
| EU220430.1 | - | 3 | 668 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tprg1l| Gene Symbol: Tprg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110318.2 | NP_001103788.1 | 3 | 668 | ||
| GenBank mRNA | BC160865.1 | - | 3 | 517 | 7324838 | |
| EU220430.1 | - | 3 | 668 | |||