siRNA Details
| Gene Symbol: Dedd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748789.2 | XP_003748837.1 | 5 | 1072 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dedd2| Gene Symbol: Dedd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748789.2 | XP_003748837.1 | 5 | 1072 | ||