siRNA Details
| Gene Symbol: LOC687119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748960.1 | XP_003749008.1 | 1 | 664 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC687119| Gene Symbol: LOC687119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748960.1 | XP_003749008.1 | 1 | 664 | ||