siRNA Details
| Gene Symbol: Zfp526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748791.3 | XP_003748839.1 | 3 | 327 | ||
| XM_006228514.2 | XP_006228576.1 | 3 | 940 | |||
| XM_006228517.2 | XP_006228579.1 | 4 | 497 | |||
| XM_006228518.2 | XP_006228580.1 | 4 | 1737 | |||
| XM_006228519.2 | XP_006228581.1 | 3 | 2666 | |||
| XM_008759065.1 | XP_008757287.1 | 4 | 1580 | |||

Zfp526
Genomic Map