siRNA Details
| Gene Symbol: Ttll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108640.2 | NP_001102110.2 | 11 | 1600 | ||
| GenBank mRNA | BC168907.1 | - | 10 | 1458 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttll3| Gene Symbol: Ttll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108640.2 | NP_001102110.2 | 11 | 1600 | ||
| GenBank mRNA | BC168907.1 | - | 10 | 1458 | ||