siRNA Details
| Gene Symbol: Fpgs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146125.2 | NP_001139597.2 | 12 | 1617 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fpgs| Gene Symbol: Fpgs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146125.2 | NP_001139597.2 | 12 | 1617 | ||