siRNA Details
| Gene Symbol: Asb17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109676.2 | NP_001103146.1 | 1 | 255 | ||
| GenBank mRNA | AJ518866.1 | - | 1 | 255 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb17| Gene Symbol: Asb17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109676.2 | NP_001103146.1 | 1 | 255 | ||
| GenBank mRNA | AJ518866.1 | - | 1 | 255 | ||