siRNA Details
| Gene Symbol: LOC691853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246833.2 | XP_006246895.1 | 1 | 1206 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC691853| Gene Symbol: LOC691853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246833.2 | XP_006246895.1 | 1 | 1206 | ||