siRNA Details
Gene Symbol: LOC691853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246833.2 | XP_006246895.1 | 3 | 1695 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC691853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246833.2 | XP_006246895.1 | 3 | 1695 |