siRNA Details
| Gene Symbol: LOC687707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145538.1 | NP_001139010.1 | 3 | 517 | ||
| GenBank mRNA | AW529560.1 | - | 2 | 182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC687707| Gene Symbol: LOC687707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145538.1 | NP_001139010.1 | 3 | 517 | ||
| GenBank mRNA | AW529560.1 | - | 2 | 182 | ||