siRNA Details
| Gene Symbol: Pip5kl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749451.3 | XP_003749499.2 | 9 | 2458 | ||
| XM_006234011.2 | XP_006234073.1 | 8 | 2404 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pip5kl1| Gene Symbol: Pip5kl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749451.3 | XP_003749499.2 | 9 | 2458 | ||
| XM_006234011.2 | XP_006234073.1 | 8 | 2404 | |||