siRNA Details
Gene Symbol: Pip5kl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749451.3 | XP_003749499.2 | 2 | 1739 | ||
XM_006234011.2 | XP_006234073.1 | 2 | 1739 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pip5kl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749451.3 | XP_003749499.2 | 2 | 1739 | ||
XM_006234011.2 | XP_006234073.1 | 2 | 1739 |