siRNA Details
| Gene Symbol: Traf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271240.1 | NP_001258169.1 | 7 | 1243 | ||
| XM_006233999.2 | XP_006234061.1 | 10 | 1600 | |||
| XM_006234000.2 | XP_006234062.1 | 10 | 1515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Traf1| Gene Symbol: Traf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271240.1 | NP_001258169.1 | 7 | 1243 | ||
| XM_006233999.2 | XP_006234061.1 | 10 | 1600 | |||
| XM_006234000.2 | XP_006234062.1 | 10 | 1515 | |||