siRNA Details
| Gene Symbol: Traf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271240.1 | NP_001258169.1 | 3 | 328 | ||
| XM_006233999.2 | XP_006234061.1 | 6 | 685 | |||
| XM_006234000.2 | XP_006234062.1 | 6 | 600 | |||
| GenBank mRNA | BC158696.1 | - | 6 | 472 | ||

Traf1
Genomic Map