siRNA Details
| Gene Symbol: Nufip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750867.3 | XP_003750915.1 | 2 | 644 | ||
| Gene Symbol: RGD1564405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749882.3 | XP_003749930.1 | 2 | 475 | ||

Nufip2
Genomic Map