siRNA Details
Gene Symbol: Tmem239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749572.3 | XP_003749620.1 | 1 | 151 | ||
XM_006235049.2 | XP_006235111.1 | 1 | 203 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749572.3 | XP_003749620.1 | 1 | 151 | ||
XM_006235049.2 | XP_006235111.1 | 1 | 203 |